missing translation for 'onlineSavingsMsg'
Learn More

PTH Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen PTH
Concentration 0.2 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898814
View Documents
Proteintech
19001-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16888814
View Documents
Proteintech
19001-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PTH is a hormone producted by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. Defects in this gene are a cause of familial isolated hypoparathyroidism.
Specifications

Specifications

PTH
Western Blot
Unconjugated
Rabbit
Human
P01270
5741
Peptide
Primary
-20°C
PTH
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Parathormone, Parathyrin, parathyroid hormone, PTH, PTH1
PTH
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PTH Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.