Learn More
Cole-Parmer™ PVDF Bulkhead Compression Union
Compression fittings are an excellent choice for connecting rigid tubing systems
Brand: Cole-Parmer™ 06390-60
This item is not returnable.
View return policy
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Very good chemical resistance
- Rated to 220psi maximum at 77°F ( 25°C)
- Usable from 32 to 212°F (0 to 100°C)
Specifications
Compression Connection | |
PVDF | |
5/16 in. | |
5/16 in. | |
11/16 in. |
Autoclavable | |
2 17/32 in. | |
5/16 in. | |
Autoclave or Ethylene Oxide | |
Bulkhead Compression Union |
Product Content Correction
Product Title
Cole-Parmer™ PVDF Bulkhead Compression Union > 5/16 in.
Spot an opportunity for improvement?Share a Content Correction