Learn More
Pyrazine, 99+% (dry wt.) water <1.0%
CAS: 290-37-9 | C4H4N2 | 80.09 g/mol
325.00 SEK - 2144.00 SEK
Chemical Identifiers
CAS 290-37-9
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006122
InChI Key KYQCOXFCLRTKLS-UHFFFAOYSA-N
Synonym 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
PubChem CID 9261
ChEBI CHEBI:30953
IUPAC Name pyrazine
SMILES C1=CN=CC=N1
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11481738
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A13982.06
5 g
325.00 SEK
5g
11491738
SDS missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
A13982.14
25 g
794.00 SEK
25g
11401748
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A13982.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Pyrazine is used in flavoring, fragrance, and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyrazine is used in flavoring, fragrance, and dyes.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : UQ2015000
missing translation for 'tsca' : TSCA
Certificates
RUO – Research Use Only
Product Content Correction
Product Title
Pyrazine, 99+% (dry wt.) water <1.0%