missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrocatechol Violet Indicator, For Metal Titration, Honeywell Fluka™
Indicator for metal titration
1186.38 SEK - 2240.20 SEK
Chemical Identifiers
CAS | 115-41-3 |
---|---|
Molecular Formula | C19H14O7S |
Molecular Weight (g/mol) | 386.37 |
MDL Number | MFCD00005868 |
InChI Key | RRRCKIRSVQAAAS-UHFFFAOYSA-N |
Synonym | Catechol violet |
PubChem CID | 66993 |
IUPAC Name | 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol |
SMILES | OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15683830
|
Honeywell-Fluka
32672-1G |
1 g | Glass Bottle |
2240.20 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15693830
|
Honeywell-Fluka
32672-5G |
5 g | Glass Bottle |
1186.38 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
115-41-3 | |
386.37 | |
RRRCKIRSVQAAAS-UHFFFAOYSA-N | |
66993 | |
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1 |
C19H14O7S | |
MFCD00005868 | |
Catechol violet | |
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diolShow More |
Specifications
185°C | |
C19H14O7S | |
NONH for all modes of transport | |
Catechol violet | |
OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1 | |
386.37 | |
386.38g/mol | |
6.5 to 8 | |
Pyrocatechol Violet |
115-41-3 | |
MFCD00005868 | |
353938 | |
RRRCKIRSVQAAAS-UHFFFAOYSA-N | |
4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol | |
66993 | |
For metal titration | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 204-088-3
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrocatechol Violet Indicator, For Metal Titration, Honeywell Fluka™