Learn More
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene Product Code.: 10451484
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Product Code. p-4812509 Shop All Thermo Scientific Chemicals Products
5 mL, Glass bottle, 5mL
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
Product Code. 10451484
Brand: Thermo Scientific Acros 369780050
Request Bulk or Custom Format
CAS: 112022-83-0 | C18H20BNO | 277.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112022-83-0, 108-88-3
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalystShow MoreShow Less
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Density 0.9500g/mL
Boiling Point 111.0°C
Flash Point 4°C
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts
Formula Weight 277.17
Chemical Name or Material (R)-(+)-2-Methyl-CBS-oxazaborolidine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452465 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number369780050,369780250
Lot NumberA0447479 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number369780050,369780250
Lot NumberA0452465 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number369780050,369780250
Lot NumberA0441743 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog Number
Lot NumberA0435672 Certificate TypeCertificate of Analysis Date07/01/2022 Catalog Number369780050,369780250
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Product Title
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene >