missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14625-1-AP-20UL
This item is not returnable.
View return policy
Description
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.Specifications
RABEP2 | |
Polyclonal | |
Unconjugated | |
RABEP2 | |
FRA, Rabaptin 5beta, RABEP2, RABPT5B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
70314, 79874, 80754 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q62835, Q91WG2, Q9H5N1 | |
Rabep2 | |
RABEP2 Fusion Protein Ag6197 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RABEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction