missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANGAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11102-1-AP-20UL
This item is not returnable.
View return policy
Description
RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1, 000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle.Specifications
RANGAP1 | |
Polyclonal | |
Unconjugated | |
RANGAP1 | |
Fug1, KIAA1835, RANGAP1, SD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19387, 362965, 5905 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Flow Cytometry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46060, P46061 | |
Rangap1 | |
RANGAP1 Fusion Protein Ag1563 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RANGAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction