Learn More
RB1 Mouse anti-Human, Unconjugated, Clone: 1A2A6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67521-1-IG-20UL
 View more versions of this product
Product Code. 16880196
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The RB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RB1. This antibody recognizes human antigen. The RB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RB1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1A2A6
Unconjugated
20 μL
5925
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RB1 Mouse anti-Human, Unconjugated, Clone: 1A2A6, Proteintech > 20 μL