missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17137-1-AP-150UL
This item is not returnable.
View return policy
Description
RBM12B gene ontology annotations related to this gene include RNA binding.Specifications
RBM12B | |
Polyclonal | |
Unconjugated | |
RBM12B | |
RBM12B, RNA binding motif protein 12B, RNA binding protein 12B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313069, 389677, 72397 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80YR9, Q8IXT5 | |
Rbm12b, Rbm12b1 | |
RBM12B Fusion Protein Ag10894 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RBM12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction