Learn More
RBM15B Mouse anti-Human, Unconjugated, Clone: 1C2C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67506-1-IG-20UL
Description
The RBM15B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBM15B. This antibody recognizes human antigen. The RBM15B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RBM15B | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1C2C11 | |
Unconjugated | |
20 μL | |
29890 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.