Learn More
RBMS2 Mouse anti-Human, Unconjugated, Clone: 1E5C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67395-1-IG-20UL
 View more versions of this product
Product Code. 16899995
  • 1645.54 SEK / 20µL
Description
The RBMS2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBMS2. This antibody recognizes human antigen. The RBMS2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RBMS2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1E5C5
Unconjugated
20 μL
5939
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RBMS2 Mouse anti-Human, Unconjugated, Clone: 1E5C5, Proteintech > 20 μL