Learn More
RBMS2 Mouse anti-Human, Unconjugated, Clone: 1E5C5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RBMS2 |
---|---|
Clone | 1E5C5 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899995
|
Proteintech
67395-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889995
|
Proteintech
67395-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RBMS2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBMS2. This antibody recognizes human antigen. The RBMS2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RBMS2 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
5939 | |
Primary |
1E5C5 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.