Learn More
RBMS2 Mouse anti-Human, Unconjugated, Clone: 1E5C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67395-1-IG-20UL
Description
The RBMS2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBMS2. This antibody recognizes human antigen. The RBMS2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RBMS2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1E5C5 | |
Unconjugated | |
20 μL | |
5939 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.