missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Retinol-binding protein 1
A DNA sequence encoding the Bos taurus (Bovine) Retinol-binding protein 3, was expressed in the hosts and tags indicated.
3490.00 SEK - 31480.00 SEK
Specifications
Accession Number | NP_776589.1 |
---|---|
Gene ID (Entrez) | 281443 |
Name | Retinol-binding protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16018191
|
enQuireBio™
QP9364-ye-10ug |
10 ÎĽg | N/A | ||||||
16058191
|
enQuireBio™
QP9364-ye-50ug |
50 ÎĽg | N/A | ||||||
16008191
|
enQuireBio™
QP9364-ye-100ug |
100 ÎĽg |
12935.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16038191
|
enQuireBio™
QP9364-ye-200ug |
200 ÎĽg | N/A | ||||||
16048191
|
enQuireBio™
QP9364-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16028191
|
enQuireBio™
QP9364-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_776589.1 | |
Retinol-binding protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281443 | |
Research Use Only | |
RBP3 | |
Recombinant Protein | |
Yeast | |
SLGELVEGTG RLLEAHYARP EVVGQMGALL RAKLAQGAYR TAVDLESLAS QLTADLQEMS GDHRLLVFHS PGEMVAEEAP PPPPVVPSPE ELSYLIEALF KTEVLPGQLG YLRFDAMAEL ETVKAVGPQL VQLVWQKLVD TAALVVDLRY NPGSYSTAVP LLCSYFFEAE PRRHLYSVFD RATSRVTEVW TLPHVTGQRY GSHKDLYVLV SHTSGSAAEA FAHTMQDLQR ATIIGEPTAG GALSVGIYQV GSSALYASMP TQMAMSASTG EAWDLAGVEP DITVPMSVAL STARDIVTLR AKVPTVLQTA GKLVADNYAS PELGVKMAAE LSGLQSRYAR VTSEAALAEL LQADLQVLSG DPHLKTAHIP EDAKDRIPGI VPMQIPSPEV FEDLIKFSFH TNVLEGNVGY LRFDMFGDCE LLTQVSELLV EHVWKKIVHT DALIVDMRFN IGGPTSSISA LCSYFFDEGP PILLDKIYNR PNNSVSELWT LSQLEGERYG SKKSMVILTS TLTAGAAEEF TYIMKRLGRA LVIGEVTSGG CQPPQTYHVD DTDLYLTIPT ARSVGAADGS SWEGVGVVPD VAVPAEAALT RAQEMLQHT | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Bovine Retinol-binding protein 1