missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.

3070.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_417389.1
Gene ID (Entrez) 947407
Name Ribose-5-phosphate isomerase A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16019070
View Documents
enQuireBio™
QP7507-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16059070
View Documents
enQuireBio™
QP7507-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16009070
View Documents
enQuireBio™
QP7507-ec-100ug
100 ÎĽg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16039070
View Documents
enQuireBio™
QP7507-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16049070
View Documents
enQuireBio™
QP7507-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16029070
View Documents
enQuireBio™
QP7507-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_417389.1
Ribose-5-phosphate isomerase A Protein
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
947407
Research Use Only
rpiA
Recombinant Protein
E. coli
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.