missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ruvC Protein
A DNA sequence encoding the Escherichia coli (strain K12) ruvC, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_416377.1 |
---|---|
Gene ID (Entrez) | 946378 |
Name | ruvC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098630
|
enQuireBio™
QP7393-ec-10ug |
10 ÎĽg | N/A | ||||||
16038640
|
enQuireBio™
QP7393-ec-50ug |
50 ÎĽg | N/A | ||||||
16088630
|
enQuireBio™
QP7393-ec-100ug |
100 ÎĽg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018640
|
enQuireBio™
QP7393-ec-200ug |
200 ÎĽg | N/A | ||||||
16028640
|
enQuireBio™
QP7393-ec-500ug |
500 ÎĽg | N/A | ||||||
16008640
|
enQuireBio™
QP7393-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_416377.1 | |
ruvC Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946378 | |
Research Use Only | |
ruvC | |
Recombinant Protein | |
E. coli | |
AIILGIDPGS RVTGYGVIRQ VGRQLSYLGS GCIRTKVDDL PSRLKLIYAG VTEIITQFQP DYFAIEQVFM AKNADSALKL GQARGVAIVA AVNQELPVFE YAARQVKQTV VGIGSAEKSQ VQHMVRTLLK LPANPQADAA DALAIAITHC HVSQNAMQMS ESRLNLARGR LR | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli ruvC Protein