missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Universal stress protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Universal stress protein F, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7202-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415894.4 | |
Universal stress protein 1 | |
Research Use Only | |
uspF | |
Recombinant Protein | |
E. coli | |
MNRTILVPID ISDSELTQRV ISHVEEEAKI DDAEVHFLTV IPSLPYYASL GLAYSAELPA MDDLKAEAKS QLEEIIKKFK LPTDRVHVHV EEGSPKDRIL ELAKKIPAHM IIIASHRPDI TTYLLGSNAA AVVRHAECSV LVVR | |
Greater than 90% as determined by SDS-PAGE. |
945948 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |