missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 2, was expressed in the hosts and tags indicated.

1640.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_001186674.1
Gene ID (Entrez) 402
Name ADP-ribosylation factor-like protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16004201
View Documents
enQuireBio™
QP8356-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16044201
View Documents
enQuireBio™
QP8356-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16094191
View Documents
enQuireBio™
QP8356-ec-100ug
100 ÎĽg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16024201
View Documents
enQuireBio™
QP8356-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16034201
View Documents
enQuireBio™
QP8356-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16014201
View Documents
enQuireBio™
QP8356-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001186674.1
ADP-ribosylation factor-like protein 1
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
402
Research Use Only
ARL2
Recombinant Protein
E. coli
LLMLGLDNAG KTTILKKFNG EDIDTISPTL GFNIKTLEHR GFKLNIWDVG GQKSLRSYWR NYFESTDGLI WVVDSADRQR MQDCQRELQS LLVEERLAGA TLLIFANKQD LPGALSSNAI REVLELDSIR SHHWCIQGCS AVTGENLLPG IDWLLDDISS RIFTAD
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.