missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6432-ec-50ug

 View more versions of this product

Product Code. 16083270

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_003986.2
Atrial natriuretic peptide receptor 2 Protein
Research Use Only
NPR2
Recombinant Protein
E. coli
RNLTLAVVLP EHNLSYAWAW PRVGPAVALA VEALGRALPV DLRFVSSELE GACSEYLAPL SAVDLKLYHD PDLLLGPGCV YPAASVARFA SHWRLPLLTA GAVASGFSAK NDHYRTLVRT GPSAPKLGEF VVTLHGHFNW TARAALLYLD ARTDDRPHYF TIEGVFEALQ GSNLSVQHQV YAREPGGPEQ ATHFIRANGR IVYICGPLEM LHEILLQAQR ENLTNGDYVF FYLDVFGESL RAGPTRATGR PWQDNRTREQ AQALREAFQT VLVITYREPP NPEYQEFQNR LLIRAREDFG VELGPSLMNL IAGCFYDGIL LYAEVLNETI QEGGTREDGL RIVEKMQGRR YHGVTGLVVM DKNNDRETDF VLWAMGDLDS GDFQPAAHYS GAEKQIWWTG RPIPWVKGAP PSDNPPCAFD LDDPSCDKTP LSTLAI
Greater than 90% as determined by SDS-PAGE.
4882
50 ÎĽg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 2 Protein > 50ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.