missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Exportin-2 Protein
A DNA sequence encoding the Homo sapiens(Human) Exportin-2, was expressed in the hosts and tags indicated.
4825.00 SEK - 20080.00 SEK
Specifications
Accession Number | NP_001243064.1 |
---|---|
Gene ID (Entrez) | 1434 |
Name | Exportin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039121
|
enQuireBio™
QP9974-iv-10ug |
10 ÎĽg | N/A | ||||||
16049121
|
enQuireBio™
QP9974-iv-50ug |
50 ÎĽg | N/A | ||||||
16029121
|
enQuireBio™
QP9974-iv-100ug |
100 ÎĽg |
20080.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001243064.1 | |
Exportin-2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1434 | |
Research Use Only | |
CSE1L | |
Recombinant Protein | |
E. coli | |
MELSDANLQT LTEYLKKTLD PDPAIRRPAE KFLESVEGNQ NYPLLLLTLL EKSQDNVIKV CASVTFKNYI KRNWRIVEDE PNKICEADRV AIKANIVHLM LSSPEQIQKQ LSDAISIIGR EDFPQKWPDL LTEMVNRFQS GDFHVINGVL RTAHSLFKRY RHEFKSNELW TEIKLVLDAF ALPLTNLFKA TIELCSTHAN | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Exportin-2 Protein