missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutaredoxin-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin-3, was expressed in the hosts and tags indicated.

1580.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_001186797.1
Gene ID (Entrez) 10539
Name Glutaredoxin-3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16020770
View Documents
enQuireBio™
QP6094-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16060770
View Documents
enQuireBio™
QP6094-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16010770
View Documents
enQuireBio™
QP6094-ec-100ug
100 ÎĽg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16040770
View Documents
enQuireBio™
QP6094-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16050770
View Documents
enQuireBio™
QP6094-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16030770
View Documents
enQuireBio™
QP6094-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001186797.1
Glutaredoxin-3 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
10539
Research Use Only
GLRX3
Recombinant Protein
E. coli
AAGAAEAAVA AVEEVGSAGQ FEELLRLKAK SLLVVHFWAP WAPQCAQMNE VMAELAKELP QVSFVKLEAE GVPEVSEKYE ISSVPTFLFF KNSQKIDRLD GAHAPELTKK VQRHASSGSF LPSANEHLKE DLNLRLKKLT HAAPCMLFMK GTPQEPRCGF SKQMVEILHK HNIQFSSFDI FSDEEVRQGL KAYSSWPTYP QLYVSGELIG GLDIIKELEA SEELDTICPK APKLEERLKV LTNKASVMLF MKGNKQEAKC GFSKQILEIL NSTGVEYETF DILEDEEVRQ GLKAYSNWPT YPQLYVKGEL VGGLDIVKEL KENGELLPIL RGEN
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Glutaredoxin-3 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.