missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-F Protein

A cDNA sequence encoding the HLA-F was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12277-100ug

 View more versions of this product

Product Code. 15999908

  • 20560.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

3134
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGSHSLRY FSTAVSRPGR GEPRYIAVEY VDDTQFLRFD SDAAIPRMEP REPWVEQEGP QYWEWTTGYA KANAQTDRVA LRNLLRRYNQ SEAGSHTLQG MNGCDMGPDG RLLRGYHQHA YDGKDYISLN EDLRSWTAAD TVAQITQRFY EAEEYAEEFR TYLEGECLEL LRRYLENGKE TLQRADPPKA HVAHHPISDH EATLRCWALG FYPAEITLTW QRDGEEQTQD TELVETRPAG DGTFQKWAAV VVPPGEEQRY TCHVQHEGLP QPLILRWEQS PQPTIPI
Greater than 90.0% as determined by SDS-PAGE.
HLA-F Protein
Research Use Only
HLA-F
Human
His
HLA-F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human HLA-F Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.