missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HPR Protein

A cDNA sequence encoding the HPR was constructed and used to recombinantly synthesize the protein.

1615.00 SEK - 92910.00 SEK

Specifications

Name HPR Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15999988
View Documents
enQuireBio™
QP12303-2ug
2 ÎĽg
1615.00 SEK
2µg
Please to purchase this item. Need a web account? Register with us today!
15979988
View Documents
enQuireBio™
QP12303-10ug
10 ÎĽg
2515.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
15989988
View Documents
enQuireBio™
QP12303-1mg
1 mg
92910.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

HPR Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HPR protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 20% glycerol 1mM DTT and 0.15M NaCl.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLYSGNDV TDISDDRFPK PPEIANGYVE HLFRYQCKNY YRLRTEGDGV YTLNDKKQWI NKAVGDKLPE CEAVCGKPKN PANPVQRILG GHLDAKGSFP WQAKMVSHHN LTTGATLINE QWLLTTAKNL FLNHSENATA KDIAPTLTLY VGKKQLVEIE KVVLHPNYHQ VDIGLIKLKQ KVLVNERVMP ICLPSKNYAE VGRVGYVSGW GQSDNFKLTD HLKYVMLPVA DQYDCITHYE GSTCPKWKAP KSPVGVQPIL NEHTFCVGMS KYQEDTCYGD AGSAFAVHDL EEDTWYAAGI LSFDKSCAVA EYGVYVKVTS IQHWVQKTIA EN
Greater than 85% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human HPR Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.