missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Inosine-5'-monophosphate dehydrogenase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Inosine-5'-monophosphate dehydrogenase 2, was expressed in the hosts and tags indicated.

1580.00 SEK - 22640.00 SEK

Specifications

Accession Number NP_000875.2
Gene ID (Entrez) 3615
Name Inosine-5'-monophosphate dehydrogenase 2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16014181
View Documents Promotion Details
enQuireBio™
QP8353-EC-100UG
100 μg
4960.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 13-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16034181
View Documents Promotion Details
enQuireBio™
QP8353-EC-1MG
1 mg
22640.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 13-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16024181
View Documents Promotion Details
enQuireBio™
QP8353-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16044181
View Documents Promotion Details
enQuireBio™
QP8353-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16054181
View Documents Promotion Details
enQuireBio™
QP8353-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16064181
View Documents Promotion Details
enQuireBio™
QP8353-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000875.2
Inosine-5'-monophosphate dehydrogenase 2 Protein
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
3615
Research Use Only
IMPD2
Recombinant Protein
E. coli
LISGGTSYVP DDGLTAQQLF NCGDGLTYND FLILPGYIDF TADQVDLTSA LTKKITLKTP LVSSPMDTVT EAGMAIAMAL TGGIGFIHHN CTPEFQANEV RKVKKYEQGF ITDPVVLSPK DRVRDVFEAK ARHGFCGIPI TDTGRMGSRL VGIISSRDID FLKEEEHDCF LEEIMTKRED LVVAPAGITL KEANEILQRS KKGKLPIVNE DDELVAIIAR TDLKKNRDYP LASKDAKKQL LCGAAIGTHE DDKYRLDLLA QAGVDVVVLD SSQGNSIFQI NMIKYIKDKY PNLQVIGGNV VTAAQAKNLI DAGVDALRVG MGSGSICITQ EVLACGRPQA TAVYKVSEYA RRFGVPVIAD GGIQNVGHIA KALALGASTV MMGSLLAATT EAPGEYFFSD GIRLKKYRGM GSLDAMDKHL SSQNRYFSEA DKIKVAQGVS GAVQDKGSIH KFVPYLIAGI QHSCQDIGAK SLTQVRAMMY SGELKFEKRT SSAQVEGGVH SLHSYEKRLF
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications