Learn More
enQuireBio™ Recombinant Human Methionine synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Methionine synthase, was expressed in the hosts and tags indicated.
2075.00 SEK - 32580.00 SEK
Specifications
Accession Number | NP_000245.2 |
---|---|
Gene ID (Entrez) | 4548 |
Name | Methionine synthase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032251
|
enQuireBio™
QP8028-ye-10ug |
10 μg | N/A | ||||||
16072251
|
enQuireBio™
QP8028-ye-50ug |
50 μg | N/A | ||||||
16022251
|
enQuireBio™
QP8028-ye-100ug |
100 μg |
7570.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16052251
|
enQuireBio™
QP8028-ye-200ug |
200 μg | N/A | ||||||
16062251
|
enQuireBio™
QP8028-ye-500ug |
500 μg | N/A | ||||||
16042251
|
enQuireBio™
QP8028-ye-1mg |
1 mg |
32580.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000245.2 | |
Methionine synthase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4548 | |
Research Use Only | |
MTR | |
Recombinant Protein | |
Yeast | |
SLKERRYLPL SQARKSGFQM DWLSEPHPVK PTFIGTQVFE DYDLQKLVDY IDWKPFFDVW QLRGKYPNRG FPKIFNDKTV GGEARKVYDD AHNMLNTLIS QKKLRARGVV GFWPAQSIQD DIHLYAEAAV PQAAEPIATF YGLRQQAEKD SASTEPYYCL SDFIAPLHSG IRDYLGLFAV ACFGVEELSK AYEDDGDDYS SIMVKALGDR LAEAFAEELH ERVRRELWAY CGSEQLDVAD LRRLRYKGIR PAPGYPSQPD HTEKLTMWRL ADIEQSTGIR LTESLAMAPA SAVSGLYFSN LKSKYFAVGK ISKDQVEDYA LRKNISVAEV EKWLGPILGY DTD | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Methionine synthase Protein