missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein
A DNA sequence encoding the Homo sapiens (Human) mRNA-decapping enzyme 1A, was expressed in the hosts and tags indicated.
2515.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_001277133.1 |
---|---|
Gene ID (Entrez) | 55802 |
Name | mRNA-decapping enzyme 1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033241
|
enQuireBio™
QP8201-ec-10ug |
10 ÎĽg | N/A | ||||||
16073241
|
enQuireBio™
QP8201-ec-50ug |
50 ÎĽg | N/A | ||||||
16023241
|
enQuireBio™
QP8201-ec-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16053241
|
enQuireBio™
QP8201-ec-200ug |
200 ÎĽg | N/A | ||||||
16063241
|
enQuireBio™
QP8201-ec-500ug |
500 ÎĽg | N/A | ||||||
16043241
|
enQuireBio™
QP8201-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001277133.1 | |
mRNA-decapping enzyme 1A Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55802 | |
Research Use Only | |
DCP1A | |
Recombinant Protein | |
E. coli | |
MEALSRAGQE MSLAALKQHD PYITSIADLT GQVALYTFCP KANQWEKTDI EGTLFVYRRS ASPYHGFTIV NRLNMHNLVE PVNKDLEFQL HEPFLLYRNA SLSIYSIWFY DKNDCHRIAK LMADVVEEET RRSQQAARDK QSPSQANGCS DHRPIDILEM LSRAKDEYER NQMGDSNISS PGLQPSTQLS NLGSTETLEE MPSGSQDKSA PSGHKHLTVE ELFGTSLPKE QPAVVGLDSE EMERLPGDAS QKEPNSFLPF PFEQLGGAPQ SETLGVPSAA HHSVQPEITT PVLITPASIT QSNEKHAPTY TIPLSPVLSP TLPAEAPTAQ VPPSLPRNST MMQAVKTTPR QRSPLLNQPV PELSHASLIA NQSPFRAPLN VTNTAGTSLP SVDLLQKLRL TPQHDQIQTQ PLGKGAMVAS FSPAAGQLAT PESFIEPPSK TAAARVAASA SLSNMVLAPL QSMQQNQDPE VFVQPKVLSS AIQVAGAPLV TATTTAVSSV LLAPSVFQQT VTRSSDLERK ASSPSPLTIG TPESQRKPSI ILSKSQLQDT LIHLIKNDSS FLSTLHEVYL QVLTKNKDNH NL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1A Protein