missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Obg-like ATPase 1, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_001011708.1 |
---|---|
Gene ID (Entrez) | 29789 |
Name | Obg-like ATPase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093400
|
enQuireBio™
QP6451-ec-10ug |
10 ÎĽg | N/A | ||||||
16033410
|
enQuireBio™
QP6451-ec-50ug |
50 ÎĽg | N/A | ||||||
16083400
|
enQuireBio™
QP6451-ec-100ug |
100 ÎĽg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013410
|
enQuireBio™
QP6451-ec-200ug |
200 ÎĽg | N/A | ||||||
16023410
|
enQuireBio™
QP6451-ec-500ug |
500 ÎĽg | N/A | ||||||
16003410
|
enQuireBio™
QP6451-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001011708.1 | |
Obg-like ATPase 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29789 | |
Research Use Only | |
OLA1 | |
Recombinant Protein | |
E. coli | |
MPPKKGGDGI KPPPIIGRFG TSLKIGIVGL PNVGKSTFFN VLTNSQASAE NFPFCTIDPN ESRVPVPDER FDFLCQYHKP ASKIPAFLNV VDIAGLVKGA HNGQGLGNAF LSHISACDGI FHLTRAFEDD DITHVEGSVD PIRDIEIIHE ELQLKDEEMI GPIIDKLEKV AVRGGDKKLK PEYDIMCKVK SWVIDQKKPV RFYHDWNDKE IEVLNKHLFL TSKPMVYLVN LSEKDYIRKK NKWLIKIKEW VDKYDPGALV IPFSGALELK LQELSAEERQ KYLEANMTQS ALPKIIKAGF AALQLEYFFT AGPDEVRAWT IRKGTKAPQA AGKIHTDFEK GFIMAEVMKY EDFKEEGSEN AVKAAGKYRQ QGRNYIVEDG DIIFFKFNTP QQPKKK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Obg-like ATPase 1 Protein