missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein

A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8259-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033611

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_055136.1
Origin recognition complex subunit 6 Protein
Research Use Only
ORC6
Recombinant Protein
E. coli
MGSELIGRLA PRLGLAEPDM LRKAEEYLRL SRVKCVGLSA RTTETSSAVM CLDLAASWMK CPLDRAYLIK LSGLNKETYQ SCLKSFECLL GLNSNIGIRD LAVQFSCIEA VNMASKILKS YESSLPQTQQ VDLDLSRPLF TSAALLSACK ILKLKVDKNK MVATSGVKKA IFDRLCKQLE KIGQQVDREP GDVATPPRKR KKIVVEAPAK EMEKVEEMPH KPQKDEDLTQ DYEEWKRKIL ENAASAQKAT AE
Greater than 80% as determined by SDS-PAGE.
23594
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers