missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 6, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_055136.1 |
---|---|
Gene ID (Entrez) | 23594 |
Name | Origin recognition complex subunit 6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093601
|
enQuireBio™
QP8259-ec-10ug |
10 ÎĽg | N/A | ||||||
16033611
|
enQuireBio™
QP8259-ec-50ug |
50 ÎĽg | N/A | ||||||
16083601
|
enQuireBio™
QP8259-ec-100ug |
100 ÎĽg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013611
|
enQuireBio™
QP8259-ec-200ug |
200 ÎĽg | N/A | ||||||
16023611
|
enQuireBio™
QP8259-ec-500ug |
500 ÎĽg | N/A | ||||||
16003611
|
enQuireBio™
QP8259-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_055136.1 | |
Origin recognition complex subunit 6 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
23594 | |
Research Use Only | |
ORC6 | |
Recombinant Protein | |
E. coli | |
MGSELIGRLA PRLGLAEPDM LRKAEEYLRL SRVKCVGLSA RTTETSSAVM CLDLAASWMK CPLDRAYLIK LSGLNKETYQ SCLKSFECLL GLNSNIGIRD LAVQFSCIEA VNMASKILKS YESSLPQTQQ VDLDLSRPLF TSAALLSACK ILKLKVDKNK MVATSGVKKA IFDRLCKQLE KIGQQVDREP GDVATPPRKR KKIVVEAPAK EMEKVEEMPH KPQKDEDLTQ DYEEWKRKIL ENAASAQKAT AE | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein