Learn More
enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8259-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055136.1 | |
Origin recognition complex subunit 6 Protein | |
Research Use Only | |
ORC6 | |
Recombinant Protein | |
E. coli | |
MGSELIGRLA PRLGLAEPDM LRKAEEYLRL SRVKCVGLSA RTTETSSAVM CLDLAASWMK CPLDRAYLIK LSGLNKETYQ SCLKSFECLL GLNSNIGIRD LAVQFSCIEA VNMASKILKS YESSLPQTQQ VDLDLSRPLF TSAALLSACK ILKLKVDKNK MVATSGVKKA IFDRLCKQLE KIGQQVDREP GDVATPPRKR KKIVVEAPAK EMEKVEEMPH KPQKDEDLTQ DYEEWKRKIL ENAASAQKAT AE | |
Greater than 80% as determined by SDS-PAGE. |
23594 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |