missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein

A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8259-ec-10ug

 View more versions of this product

Product Code. 16093601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_055136.1
Origin recognition complex subunit 6 Protein
Research Use Only
ORC6
Recombinant Protein
E. coli
MGSELIGRLA PRLGLAEPDM LRKAEEYLRL SRVKCVGLSA RTTETSSAVM CLDLAASWMK CPLDRAYLIK LSGLNKETYQ SCLKSFECLL GLNSNIGIRD LAVQFSCIEA VNMASKILKS YESSLPQTQQ VDLDLSRPLF TSAALLSACK ILKLKVDKNK MVATSGVKKA IFDRLCKQLE KIGQQVDREP GDVATPPRKR KKIVVEAPAK EMEKVEEMPH KPQKDEDLTQ DYEEWKRKIL ENAASAQKAT AE
Greater than 80% as determined by SDS-PAGE.
23594
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Origin recognition complex subunit 6 Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.