Learn More
enQuireBio™ Recombinant Human Protein E7 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Name | Protein E7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | E7 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098480
|
enQuireBio™
QP7367-ye-10ug |
10 ÎĽg | N/A | ||||||
16038490
|
enQuireBio™
QP7367-ye-50ug |
50 ÎĽg | N/A | ||||||
16088480
|
enQuireBio™
QP7367-ye-100ug |
100 ÎĽg |
7570.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018490
|
enQuireBio™
QP7367-ye-200ug |
200 ÎĽg | N/A | ||||||
16028490
|
enQuireBio™
QP7367-ye-500ug |
500 ÎĽg |
20530.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16008490
|
enQuireBio™
QP7367-ye-1mg |
1 mg | N/A | ||||||
Specifications
Protein E7 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
E7 | |
Recombinant Protein | |
Yeast | |
MHGPKATLQD IVLHLEPQNE IPVDLLCHEQ LSDSEEENDE IDGVNHQHLP ARRAEPQRHT MLCMCCKCEA RIKLVVESSA DDLRAFQQLF LNTLSFVCPW CASQQ | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Human Protein E7 Protein