missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TNF-alpha Hyperactive Mutant Protein
Mutant form with approximately 5-fold greater activity
3105.00 SEK - 4525.00 SEK
Specifications
Accession Number | P01375 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 7124 |
Molecular Weight (g/mol) | 17 kDa |
Name | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757576
|
R&D Systems™
8599-TA-025/CF |
25 ÎĽg |
3105.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747766
|
R&D Systems™
8599-TA-100/CF |
100 ÎĽg |
4525.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Measured in a cytotoxicity assay using L-929 mouse fibroblast cells in the presence of the metabolic inhibitor actinomycin D.
- ED50 for this effect is 20 to 100 pg/mL.
Specifications
P01375 | |
7124 | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 100 ÎĽg/mL in PBS. | |
<0.10 EU per 1 ÎĽg of the protein by the LAL method. |
Bioactivity | |
17 kDa | |
E. coli-derived human TNF-alpha protein Lys87-Leu233 (Leu233Phe), with an N-terminal Met-Arg-Lys-Arg | |
TNF | |
Human | |
Lyophilized from a 0.2 ÎĽm filtered solution in PBS. | |
>95%, by SDS-PAGE with silver staining. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human TNF-alpha Hyperactive Mutant Protein