missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription termination factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription termination factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8725-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001192225.1 | |
Transcription termination factor 1 Protein | |
Research Use Only | |
TTF1 | |
Recombinant Protein | |
E. coli | |
HTPVSDKKKK KCSIHKERPQ KHSHEIFRDS SLVNEQSQIT RRKKRKKDFQ HLISSPLKKS RICDETANAT STLKKRKKRR YSALEVDEEA GVTVVLVDKE NINNTPKHFR KDVDVVCVDM SIEQKLPRKP KTDKFQVLAK SHAHKSEALH SKVREKKNKK HQRKAASWES QRARDTLPQS ESHQEESWLS VGPGGEITEL PASAHKNKSK KKKKKSSNRE YETLAMPEGS QA | |
Greater than 90% as determined by SDS-PAGE. |
7270 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Transcription termination factor 1 Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction