missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription termination factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription termination factor 1, was expressed in the hosts and tags indicated.

1640.00 SEK - 22640.00 SEK

Specifications

Accession Number NP_001192225.1
Gene ID (Entrez) 7270
Name Transcription termination factor 1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16006151
View Documents Promotion Details
enQuireBio™
QP8725-EC-100UG
100 μg
4960.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16026151
View Documents Promotion Details
enQuireBio™
QP8725-EC-1MG
1 mg
22640.00 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 09-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16016151
View Documents Promotion Details
enQuireBio™
QP8725-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16036151
View Documents Promotion Details
enQuireBio™
QP8725-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16046151
View Documents Promotion Details
enQuireBio™
QP8725-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16056151
View Documents Promotion Details
enQuireBio™
QP8725-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001192225.1
Transcription termination factor 1 Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7270
Research Use Only
TTF1
Recombinant Protein
E. coli
HTPVSDKKKK KCSIHKERPQ KHSHEIFRDS SLVNEQSQIT RRKKRKKDFQ HLISSPLKKS RICDETANAT STLKKRKKRR YSALEVDEEA GVTVVLVDKE NINNTPKHFR KDVDVVCVDM SIEQKLPRKP KTDKFQVLAK SHAHKSEALH SKVREKKNKK HQRKAASWES QRARDTLPQS ESHQEESWLS VGPGGEITEL PASAHKNKSK KKKKKSSNRE YETLAMPEGS QA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications