missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 Q2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7927-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001138807.1 | |
Ubiquitin-conjugating enzyme E2 Q2 Protein | |
Research Use Only | |
UBE2Q2 | |
Recombinant Protein | |
E. coli | |
MSVSGLKAEL KFLASIFDKN HERFRIVSWK LDELHCQFLV PQQGSPHSLP PPLTLHCNIT ESYPSSSPIW FVDSEDPNLT SVLERLEDTK NNNLLRQQLK WLICELCSLY NLPKHLDVEM LDQPLPTGQN GTTEEVTSEE EEEEEEMAED IEDLDHYEMK EEEPISGKKS EDEGIEKENL AILEKIRKTQ RQDHLNGAVS GSVQASDRLM KELRDIYRSQ SYKTGIYSVE LINDSLYDWH VKLQKVDPDS PLHSDLQILK EKEGIEYILL NFSFKDNFPF DPPFVRVVLP VLSGGYVLGG GALCMELLTK QGWSSAYSIE SVIMQINATL VKGKARVQFG ANKNQYNLAR AQQSYNSIVQ IHEKNGWYTP PKEDG | |
Greater than 90% as determined by SDS-PAGE. |
92912 | |
50 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction