missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Hemagglutinin-esterase-fusion glycoProtein
A DNA sequence encoding the Influenza C virus (strain C/California/1978) Hemagglutinin-esterase-fusion glycoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9694-ye-500ug
This item is not returnable.
View return policy
Specifications
Influenza Hemagglutinin-esterase-fusion glycoProtein | |
Research Use Only | |
HE | |
Recombinant Protein | |
Yeast | |
EKIKICLQKQ VNSSFSLHNG FGGNLYATEE KRMFELVKPK AGASVLNQST WIGFGDSRTD QSNSAFPRSL MSAKTADKFR SLSGGSLMLS MFGPPGKVDY LYQGCGKHKV FYEGVNWSPH AAIDCYRKNW TDIKLNFQKS IYELASQSHC MSLVNALDKT IPLQVTKGVA KNCNNSFLKN PALYTQEVKP LEQICGEENL AFFTLPTQFG TYECKLHLVA SCYFIYDSKE VYNKRGCGNY FQVIYDSSGK VVGGLDNRVS PYTGNSGDTP TMQCDMLQLK PGRYSVRSSP RFLLMPERSY CFDMKEKGPV TAVQSIWGKG RKSDYAVDQA CLSTPGCMLI QKQKPYIGEA DDHHGDQEMR ELLSGLDYEA RCISQSGWVN ETSPFTEEYL LPPKFGRCPL AAKEESIPKI PDGLLIPTSG TDTTVTKPKS RIFGIDDLII GLLFVAIVEA GIGGYLLGSR KESGGGVTKE SAEKGFEKIG NDIQILRSST NIAIEKLNDR ISHDEQAIRD LTLEIENARS EALLGELGII RALLVGNISI GLQESLWELA SEITNRAGDL AVEVSPGCWI IDNNICDQSC QNFIFKFNET APVPTIPPLD TKIDLQSDPF YWGSS | |
Greater than 90% as determined by SDS-PAGE. |
500 ÎĽg | |
Not determined. | |
Extracellular Domain | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Influenza Hemagglutinin-esterase-fusion glycoProtein > 500μg
Spot an opportunity for improvement?Share a Content Correction