missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Hemagglutinin-esterase-fusion glycoProtein
A DNA sequence encoding the Influenza C virus (strain C/California/1978) Hemagglutinin-esterase-fusion glycoprotein, was expressed in the hosts and tags indicated.
3490.00 SEK - 32610.00 SEK
Specifications
Name | Influenza Hemagglutinin-esterase-fusion glycoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HE |
Protein Length | Extracellular Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078581
|
enQuireBio™
QP9694-ye-10ug |
10 ÎĽg | N/A | ||||||
16018591
|
enQuireBio™
QP9694-ye-50ug |
50 ÎĽg | N/A | ||||||
16068581
|
enQuireBio™
QP9694-ye-100ug |
100 ÎĽg |
12935.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16098581
|
enQuireBio™
QP9694-ye-200ug |
200 ÎĽg | N/A | ||||||
16008591
|
enQuireBio™
QP9694-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16088581
|
enQuireBio™
QP9694-ye-1mg |
1 mg | N/A | ||||||
Specifications
Influenza Hemagglutinin-esterase-fusion glycoProtein | |
Not determined. | |
Extracellular Domain | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HE | |
Recombinant Protein | |
Yeast | |
EKIKICLQKQ VNSSFSLHNG FGGNLYATEE KRMFELVKPK AGASVLNQST WIGFGDSRTD QSNSAFPRSL MSAKTADKFR SLSGGSLMLS MFGPPGKVDY LYQGCGKHKV FYEGVNWSPH AAIDCYRKNW TDIKLNFQKS IYELASQSHC MSLVNALDKT IPLQVTKGVA KNCNNSFLKN PALYTQEVKP LEQICGEENL AFFTLPTQFG TYECKLHLVA SCYFIYDSKE VYNKRGCGNY FQVIYDSSGK VVGGLDNRVS PYTGNSGDTP TMQCDMLQLK PGRYSVRSSP RFLLMPERSY CFDMKEKGPV TAVQSIWGKG RKSDYAVDQA CLSTPGCMLI QKQKPYIGEA DDHHGDQEMR ELLSGLDYEA RCISQSGWVN ETSPFTEEYL LPPKFGRCPL AAKEESIPKI PDGLLIPTSG TDTTVTKPKS RIFGIDDLII GLLFVAIVEA GIGGYLLGSR KESGGGVTKE SAEKGFEKIG NDIQILRSST NIAIEKLNDR ISHDEQAIRD LTLEIENARS EALLGELGII RALLVGNISI GLQESLWELA SEITNRAGDL AVEVSPGCWI IDNNICDQSC QNFIFKFNET APVPTIPPLD TKIDLQSDPF YWGSS | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Influenza Hemagglutinin-esterase-fusion glycoProtein