missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ADH7 Protein

A DNA sequence encoding the Mus musculus (Mouse) ADH7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7693-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16000231

  • 32325.00 SEK / 1mg
Estimated Shipment: 12-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033756.2
ADH7 Protein
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
11529
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADH7
Recombinant Protein
E. coli
MGTAGKVIKC KAAVLWGVNQ PFSIEEIEVA PPKAKEVRVK ILATGICRTD DHIIKGSMVS KFPVIVGHEA VGVVESVGEG VTTVRPGDKV IPLFLPQCRE CNACLNPEGN LCIRSDLTGR GVLADGTTRF TCKGKPVQHF MNTSTFTEYT VLDESSVAKV DGAAPPEKAC LIGCGFSTGY GAAVKTAKVT PGSTCVVFGL GGVGLSVIMG CKAAGASRII GIDINKDKFQ KALAVGATEC ISPKDSTKPI SEVLSDMTGN TIQYTFEVIG RLETMVDALS SCHMNYGTSV VVGAPPSAKM LTYDPMLLFT GRTWKGCVFG GWKSRDDVPK LVTEFLEKKF DLDQLITHTL PFNNINEGFE LLYSGKSIRT VLTF
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers