missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein

A DNA sequence encoding the Mus musculus (Mouse) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7956-ye-10ug

 View more versions of this product

Product Code. 16001831

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001258282.1
Cystathionine beta-synthase Protein
Research Use Only
CBS
Recombinant Protein
Yeast
PSGTSQCEDG SAGGFQHLDM HSEKRQLEKG PSGDKDRVWI RPDTPSRCTW QLGRAMADSP HYHTVLTKSP KILPDILRKI GNTPMVRINK ISKNAGLKCE LLAKCEFFNA GGSVKDRISL RMIEDAERAG NLKPGDTIIE PTSGNTGIGL ALAAAVKGYR CIIVMPEKMS MEKVDVLRAL GAEIVRTPTN ARFDSPESHV GVAWRLKNEI PNSHILDQYR NASNPLAHYD DTAEEILQQC DGKLDMLVAS AGTGGTITGI ARKLKEKCPG CKIIGVDPEG SILAEPEELN QTEQTAYEVE GIGYDFIPTV LDRAVVDKWF KSNDEDSFAF ARMLIAQEGL LCGGSSGSAM AVAVKAAREL QEGQRCVVIL PDSVRNYMSK FLSDKWMLQK GFMKEELSVK RPWWWRLRVQ ELSLSAPLTV LPTVTCEDTI AILREKGFDQ APVVNESGAI LGMVTLGNML SSLLAGKVRP SDEVCKVLYK QFKPIHLTDT LGTLSHILEM DHFALVVHEQ IQSRDQAWSG VVGGPTDCSN GMSSKQQMVF GVVTAIDLLN FVAAREQTQT
Greater than 90% as determined by SDS-PAGE.
12411
10 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.