missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein
A DNA sequence encoding the Mus musculus (Mouse) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.
2850.00 SEK - 48075.00 SEK
Specifications
Accession Number | NP_001258282.1 |
---|---|
Gene ID (Entrez) | 12411 |
Name | Cystathionine beta-synthase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16001831
|
enQuireBio™
QP7956-ye-10ug |
10 ÎĽg | N/A | ||||||
16041831
|
enQuireBio™
QP7956-ye-50ug |
50 ÎĽg | N/A | ||||||
16091821
|
enQuireBio™
QP7956-ye-100ug |
100 ÎĽg |
10850.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16021831
|
enQuireBio™
QP7956-ye-200ug |
200 ÎĽg | N/A | ||||||
16031831
|
enQuireBio™
QP7956-ye-500ug |
500 ÎĽg | N/A | ||||||
16011831
|
enQuireBio™
QP7956-ye-1mg |
1 mg |
48075.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001258282.1 | |
Cystathionine beta-synthase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12411 | |
Research Use Only | |
CBS | |
Recombinant Protein | |
Yeast | |
PSGTSQCEDG SAGGFQHLDM HSEKRQLEKG PSGDKDRVWI RPDTPSRCTW QLGRAMADSP HYHTVLTKSP KILPDILRKI GNTPMVRINK ISKNAGLKCE LLAKCEFFNA GGSVKDRISL RMIEDAERAG NLKPGDTIIE PTSGNTGIGL ALAAAVKGYR CIIVMPEKMS MEKVDVLRAL GAEIVRTPTN ARFDSPESHV GVAWRLKNEI PNSHILDQYR NASNPLAHYD DTAEEILQQC DGKLDMLVAS AGTGGTITGI ARKLKEKCPG CKIIGVDPEG SILAEPEELN QTEQTAYEVE GIGYDFIPTV LDRAVVDKWF KSNDEDSFAF ARMLIAQEGL LCGGSSGSAM AVAVKAAREL QEGQRCVVIL PDSVRNYMSK FLSDKWMLQK GFMKEELSVK RPWWWRLRVQ ELSLSAPLTV LPTVTCEDTI AILREKGFDQ APVVNESGAI LGMVTLGNML SSLLAGKVRP SDEVCKVLYK QFKPIHLTDT LGTLSHILEM DHFALVVHEQ IQSRDQAWSG VVGGPTDCSN GMSSKQQMVF GVVTAIDLLN FVAAREQTQT | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Cystathionine beta-synthase Protein