missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hyaluronan synthase 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hyaluronan synthase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6144-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_032242.3 | |
Hyaluronan synthase 2 Protein | |
Research Use Only | |
Has2 | |
Recombinant Protein | |
Yeast | |
EHRKMKKSLE TPIKLNKTVA LCIAAYQEDP DYLRKCLQSV KRLTYPGIKV VMVIDGNSDD DLYMMDIFSE VMGRDKSATY IWKNNFHEKG PGETEESHKE SSQHVTQLVL SNKSICIMQK WGGKREVMYT AFRALGRSVD YVQVCDSDTM LDPASSVEMV KVLEEDPMVG GVGGDVQILN KYDSWISFLS SVRYWMAFNI ERACQSYFGC VQCISGPLGM YRNSLLHEFV EDWYNQEFMG NQCSFGDDRH LTNRVLSLGY ATKYTARSKC LTETPIEYLR WLNQQTRWSK SYFREWLYNA MWFHKHHL | |
Greater than 90% as determined by SDS-PAGE. |
15117 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Hyaluronan synthase 2 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction