missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hyaluronan synthase 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Hyaluronan synthase 2, was expressed in the hosts and tags indicated.
2515.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_032242.3 |
---|---|
Gene ID (Entrez) | 15117 |
Name | Hyaluronan synthase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081130
|
enQuireBio™
QP6144-ye-10ug |
10 ÎĽg | N/A | ||||||
16021140
|
enQuireBio™
QP6144-ye-50ug |
50 ÎĽg | N/A | ||||||
16071130
|
enQuireBio™
QP6144-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16001140
|
enQuireBio™
QP6144-ye-200ug |
200 ÎĽg | N/A | ||||||
16011140
|
enQuireBio™
QP6144-ye-500ug |
500 ÎĽg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16091130
|
enQuireBio™
QP6144-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_032242.3 | |
Hyaluronan synthase 2 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15117 | |
Research Use Only | |
Has2 | |
Recombinant Protein | |
Yeast | |
EHRKMKKSLE TPIKLNKTVA LCIAAYQEDP DYLRKCLQSV KRLTYPGIKV VMVIDGNSDD DLYMMDIFSE VMGRDKSATY IWKNNFHEKG PGETEESHKE SSQHVTQLVL SNKSICIMQK WGGKREVMYT AFRALGRSVD YVQVCDSDTM LDPASSVEMV KVLEEDPMVG GVGGDVQILN KYDSWISFLS SVRYWMAFNI ERACQSYFGC VQCISGPLGM YRNSLLHEFV EDWYNQEFMG NQCSFGDDRH LTNRVLSLGY ATKYTARSKC LTETPIEYLR WLNQQTRWSK SYFREWLYNA MWFHKHHL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Hyaluronan synthase 2 Protein