missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Proto-oncogene Wnt-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6887-ec-50ug

 View more versions of this product

Product Code. 16086560

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_033547.1
Proto-oncogene Wnt-3 Protein
Research Use Only
Wnt3
Recombinant Protein
E. coli
GYPIWWSLAL GQQYTSLASQ PLLCGSIPGL VPKQLRFCRN YIEIMPSVAE GVKLGIQECQ HQFRGRRWNC TTIDDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTSTICGC DSHHKGPPGE GWKWGGCSED ADFGVLVSRE FADARENRPD ARSAMNKHNN EAGRTTILDH MHLKCKCHGL SGSCEVKTCW WAQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRA KYALFKPPTE RDLVYYENSP NFCEPNPETG SFGTRDRTCN VTSHGIDGCD LLCCGRGHNT RTEKRKEKCH CVFHWCCYVS CQECIRIYDV HTCK
Greater than 90% as determined by SDS-PAGE.
22415
50 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein > 50μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.