missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Proto-oncogene Wnt-3, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_033547.1 |
---|---|
Gene ID (Entrez) | 22415 |
Name | Proto-oncogene Wnt-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046560
|
enQuireBio™
QP6887-ec-10ug |
10 μg | N/A | ||||||
16086560
|
enQuireBio™
QP6887-ec-50ug |
50 μg | N/A | ||||||
16036560
|
enQuireBio™
QP6887-ec-100ug |
100 μg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16066560
|
enQuireBio™
QP6887-ec-200ug |
200 μg | N/A | ||||||
16076560
|
enQuireBio™
QP6887-ec-500ug |
500 μg | N/A | ||||||
16056560
|
enQuireBio™
QP6887-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_033547.1 | |
Proto-oncogene Wnt-3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
22415 | |
Research Use Only | |
Wnt3 | |
Recombinant Protein | |
E. coli | |
GYPIWWSLAL GQQYTSLASQ PLLCGSIPGL VPKQLRFCRN YIEIMPSVAE GVKLGIQECQ HQFRGRRWNC TTIDDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTSTICGC DSHHKGPPGE GWKWGGCSED ADFGVLVSRE FADARENRPD ARSAMNKHNN EAGRTTILDH MHLKCKCHGL SGSCEVKTCW WAQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRA KYALFKPPTE RDLVYYENSP NFCEPNPETG SFGTRDRTCN VTSHGIDGCD LLCCGRGHNT RTEKRKEKCH CVFHWCCYVS CQECIRIYDV HTCK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Proto-oncogene Wnt-3 Protein