missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ripk3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Ripk3, was expressed in the hosts and tags indicated.
2430.00 SEK - 26260.00 SEK
Specifications
Accession Number | NP_064339.2 |
---|---|
Gene ID (Entrez) | 56532 |
Name | Ripk3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099001
|
enQuireBio™
QP9919-ye-10ug |
10 ÎĽg | N/A | ||||||
16039011
|
enQuireBio™
QP9919-ye-50ug |
50 ÎĽg | N/A | ||||||
16089001
|
enQuireBio™
QP9919-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16019011
|
enQuireBio™
QP9919-ye-200ug |
200 ÎĽg | N/A | ||||||
16029011
|
enQuireBio™
QP9919-ye-500ug |
500 ÎĽg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16009011
|
enQuireBio™
QP9919-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_064339.2 | |
Ripk3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56532 | |
Research Use Only | |
Ripk3 | |
Recombinant Protein | |
Yeast | |
MSSVKLWPTG ASAVPLVSRE ELKKLEFVGK GGFGVVFRAH HRTWNHDVAV KIVNSKKISW EVKAMVNLRN ENVLLLLGVT EDLQWDFVSG QALVTRFMEN GSLAGLLQPE CPRPWPLLCR LLQEVVLGMC YLHSLDPPLL HRDLKPSNIL LDPELHAKLA DFGLSTFQGG SQSGSGSGSG SRDSGGTLAY LDPELLFKVN LKASKASDVY SFGILVWAVL AGREAELVDK TSLIRETVCD RQSRPPLTEL PPGSPETPGL EKLKELMIHC WGSQSENRPS FQDCEPKTNE VYNLVKDKVD AAVSEVKHYL SQHRSSGRNL SAREPSQRGT EMDCPRETMV SKMLDRLHLE EPSGPVPGKC PERQAQDTSV GPATPARTSS DPVAGTPQIP HTLPFRGTTP GPVFTETPGP HPQRNQGDGR HGTPWYPWTP PNPMTGPPAL VFNNCSEVQI GNYNSLVAPP RTTASSSAKY DQAQFGRGRG WQPFHK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Ripk3 Protein