missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Secretoglobin family 3A member 2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Secretoglobin family 3A member 2, was expressed in the hosts and tags indicated.

2430.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_001276572.1
Gene ID (Entrez) 117158
Name Secretoglobin family 3A member 2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16001651
View Documents
enQuireBio™
QP7929-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16041651
View Documents
enQuireBio™
QP7929-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16091641
View Documents
enQuireBio™
QP7929-ec-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16021651
View Documents
enQuireBio™
QP7929-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16031651
View Documents
enQuireBio™
QP7929-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16011651
View Documents
enQuireBio™
QP7929-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001276572.1
Secretoglobin family 3A member 2 Protein
Not determined.
Full Length
Mouse
His-GST
Tris-based buffer, 50% glycerol
117158
Research Use Only
Scgb3a2
Recombinant Protein
E. coli
LLINRLPVVD KLPVPLDDII PSFDPLKMLL KTLGISVEHL VTGLKKCVDE LGPEASEAVK KLLVIIICSY FPGRSLCYVN NLPSFVSVLF LPMICAYPRD SKKQTFAFIE RVFEQSKL
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Secretoglobin family 3A member 2 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.