missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SerpinG1 / C1 inhibitor / C1IN Protein

A DNA sequence encoding the Mus musculus (Mouse) SerpinG1 / C1 inhibitor / C1IN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6676-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005020

  • 32325.00 SEK / 1mg
Estimated Shipment: 12-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033906.2
SerpinG1 / C1 inhibitor / C1IN Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
12258
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SERPING1
Recombinant Protein
E. coli
TQDPLEAQAK SRESFPERDD SWSPPEPTVL PSTWPTTSVA ITITNDTMGK VANESFSQHS QPAAQLPTDS PGQPPLNSSS QPSTASDLPT QATTEPFCPE PLAQCSDSDR DSSEAKLSEA LTDFSVKLYH AFSATKMAKT NMAFSPFSIA SLLTQVLLGA GDSTKSNLES ILSYPKDFAC VHQALKGFSS KGVTSVSQIF HSPDLAIRDT YVNASQSLYG SSPRVLGPDS AANLELINTW VAENTNHKIR KLLDSLPSDT CLVLLNAVYL SAKWKITFEP KKMMAPFFYK NSMIKVPMMS SVKYPVAQFD DHTLKAKVGQ LQLSHNLSFV IVVPVFPKHQ LKDVEKALNP TVFKAIMKKL ELSKFLPTYL TMPHIKVKSS QDMLSVMEKL EFFDFTYDLN LCGLTEDPDL QVSAMKHETV LELTESGVEA AAASAISFGR SLPIFEVQRP FLFLLWDQQH RFPVFMGRVY DPR
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers