missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SerpinG1 / C1 inhibitor / C1IN Protein
A DNA sequence encoding the Mus musculus (Mouse) SerpinG1 / C1 inhibitor / C1IN, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_033906.2 |
---|---|
Gene ID (Entrez) | 12258 |
Name | SerpinG1 / C1 inhibitor / C1IN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095010
|
enQuireBio™
QP6676-ec-10ug |
10 ÎĽg | N/A | ||||||
16035020
|
enQuireBio™
QP6676-ec-50ug |
50 ÎĽg | N/A | ||||||
16085010
|
enQuireBio™
QP6676-ec-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16015020
|
enQuireBio™
QP6676-ec-200ug |
200 ÎĽg | N/A | ||||||
16025020
|
enQuireBio™
QP6676-ec-500ug |
500 ÎĽg | N/A | ||||||
16005020
|
enQuireBio™
QP6676-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_033906.2 | |
SerpinG1 / C1 inhibitor / C1IN Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
12258 | |
Research Use Only | |
SERPING1 | |
Recombinant Protein | |
E. coli | |
TQDPLEAQAK SRESFPERDD SWSPPEPTVL PSTWPTTSVA ITITNDTMGK VANESFSQHS QPAAQLPTDS PGQPPLNSSS QPSTASDLPT QATTEPFCPE PLAQCSDSDR DSSEAKLSEA LTDFSVKLYH AFSATKMAKT NMAFSPFSIA SLLTQVLLGA GDSTKSNLES ILSYPKDFAC VHQALKGFSS KGVTSVSQIF HSPDLAIRDT YVNASQSLYG SSPRVLGPDS AANLELINTW VAENTNHKIR KLLDSLPSDT CLVLLNAVYL SAKWKITFEP KKMMAPFFYK NSMIKVPMMS SVKYPVAQFD DHTLKAKVGQ LQLSHNLSFV IVVPVFPKHQ LKDVEKALNP TVFKAIMKKL ELSKFLPTYL TMPHIKVKSS QDMLSVMEKL EFFDFTYDLN LCGLTEDPDL QVSAMKHETV LELTESGVEA AAASAISFGR SLPIFEVQRP FLFLLWDQQH RFPVFMGRVY DPR | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse SerpinG1 / C1 inhibitor / C1IN Protein