missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SerpinG1 / C1 inhibitor / C1IN Protein

A DNA sequence encoding the Mus musculus (Mouse) SerpinG1 / C1 inhibitor / C1IN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6676-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025020

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_033906.2
SerpinG1 / C1 inhibitor / C1IN Protein
Research Use Only
SERPING1
Recombinant Protein
E. coli
TQDPLEAQAK SRESFPERDD SWSPPEPTVL PSTWPTTSVA ITITNDTMGK VANESFSQHS QPAAQLPTDS PGQPPLNSSS QPSTASDLPT QATTEPFCPE PLAQCSDSDR DSSEAKLSEA LTDFSVKLYH AFSATKMAKT NMAFSPFSIA SLLTQVLLGA GDSTKSNLES ILSYPKDFAC VHQALKGFSS KGVTSVSQIF HSPDLAIRDT YVNASQSLYG SSPRVLGPDS AANLELINTW VAENTNHKIR KLLDSLPSDT CLVLLNAVYL SAKWKITFEP KKMMAPFFYK NSMIKVPMMS SVKYPVAQFD DHTLKAKVGQ LQLSHNLSFV IVVPVFPKHQ LKDVEKALNP TVFKAIMKKL ELSKFLPTYL TMPHIKVKSS QDMLSVMEKL EFFDFTYDLN LCGLTEDPDL QVSAMKHETV LELTESGVEA AAASAISFGR SLPIFEVQRP FLFLLWDQQH RFPVFMGRVY DPR
Greater than 90% as determined by SDS-PAGE.
12258
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers